Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OS9 Recombinant Rabbit Monoclonal Antibody (JG38-07)
GREENER_CHOICE

Catalog No. PIMA534773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534773 Supplier Invitrogen™ Supplier No. MA534773
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Mouse testis tissue lysate, rat testis tissue lysate, rat epididymis tissue, human breast cancer tissue, human placenta tissue, mouse testis tissue.

OS9 is an ER glycoprotein found in the intralumenal level and highly expressed in tumor tissue. The OS9 gene is transcriptionally induced upon activation of the Ire1/Xbp1 ER-stress pathway.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OS9
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Recombinant Monoclonal
Clone JG38-07
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene OS9
Gene Accession No. Q13438
Gene Alias 4632413K17Rik; amplified in osteosarcoma; amplified in osteosarcoma 9; AU022351; endoplasmic reticulum lectin 2; ERLEC2; erlectin 2; OS9; Os-9; OS9 endoplasmic reticulum lectin; OS9, endoplasmic reticulum lectin; osteosarcoma amplified 9; osteosarcoma amplified 9, endoplasmic reticulum associated protein; osteosarcoma amplified 9, endoplasmic reticulum lectin; protein OS-9
Gene Symbols OS9
Host Species Rabbit
Immunogen Recombinant protein within Human OS9 aa 561-667.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10956
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.