Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530110
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530110 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530110 Supplier Invitrogen™ Supplier No. PA530110
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji, mouse brain. Predicted reactivity: Mouse (97%), Rat (98%), Rabbit (97%), Rhesus Monkey (99%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSBP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.76 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene Osbp
Gene Accession No. P22059, Q3B7Z2
Gene Alias 1110018F06Rik; AW559088; HGNC:8503; Kiaa4220; mKIAA4220; OSBP; OSBP1; oxysterol binding protein; oxysterol-binding protein 1
Gene Symbols Osbp
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human OSBP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5007, 76303
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.