Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OSBP Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518218
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
Oxysterol binding protein is an intracellular protein that is believed to transport sterols from lysosomes to the nucleus where the sterol down-regulates the genes for the LDL receptor, HMG-CoA reductase, and HMG synthetase.
Specifications
| OSBP | |
| Polyclonal | |
| Unconjugated | |
| Osbp | |
| 1110018F06Rik; AW559088; HGNC:8503; Kiaa4220; mKIAA4220; OSBP; OSBP1; oxysterol binding protein; oxysterol-binding protein 1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5007 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P22059 | |
| Osbp | |
| Synthetic peptide sequence (CKEKQDWSSCPDIF) corresponding to the C-terminus amino acids of OSBP (aa 794-807). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction