Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSBP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554432
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554432 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554432 Supplier Invitrogen™ Supplier No. PA554432
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SRKWQRALQY EQEQRVHLEE TIEQLAKQHN SLERAFHSAP GRPANPSKSF IEGSLLTPKG EDSEEDEDTE YFDAMEDSTS FITV Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.

Oxysterols are byproducts of cholesterol that can have cytotoxic effects on many cell types. The membrane-bound protein encoded by this gene contains a pleckstrin homology (PH) domain and an oxysterol-binding region. It binds oxysterols such as 7-ketocholesterol and may inhibit their cytotoxicity. Alternate transcriptional splice variants have been observed but have not been fully characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSBP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OSBP2
Gene Accession No. Q969R2
Gene Alias 1700095P05Rik; C630001G20Rik; Gm244; HLM; KIAA1664; ORP4; ORP-4; Osbp2; OSBPL1; OSBPL4; OSBP-related protein 4; oxysterol binding protein 2; oxysterol binding protein-like 1; oxysterol-binding protein 2; Oxysterol-binding protein-related protein 4; RGD1309479
Gene Symbols OSBP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OSBP2. Recombinant protein control fragment (Product #RP-92362).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23762
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.