Learn More
OSBPL10/ORP10 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304885AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.Specifications
OSBPL10/ORP10 | |
Polyclonal | |
Unconjugated | |
OSBPL10 | |
OSBPL10; oxysterol binding protein-like 10; HGNC:16395; FLJ20363; ORP10; OSBP9; OSBP-related protein 10; oxysterol-binding protein-like protein 10; oxysterol-binding protein-related protein 10; OTTHUMP00000161257 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114884 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q9BXB5 | |
OSBPL10 | |
Between 1 and 50 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.