Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSBPL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518437
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518437 Supplier Invitrogen™ Supplier No. PA518437
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSBPL10
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene OSBPL10
Gene Accession No. Q9BXB5
Gene Alias 4933433D06Rik; C820004B04Rik; FLJ20363; HGNC:16395; OPR-10; ORP10; ORP-10; OSBP9; OSBPL10; OSBP-related protein 10; OTTHUMP00000161257; oxysterol binding protein like 10; oxysterol binding protein-like 10; oxysterol-binding protein-related protein 10; RGD1561652
Gene Symbols OSBPL10
Host Species Goat
Immunogen Synthetic peptide sequence (DAATEQKRHLEEKQ) corresponding to the internal amino acids of OSBPL10 (aa 715-729).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114884
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.