Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSBPL10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589070
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589070 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589070 Supplier Invitrogen™ Supplier No. PA589070
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ASPCSQRHLS VGAPGVVTIT HHKSPAAARR AKSQYSGQLH EVREMMNQVE GQQKNLVHAI ESLPGSGPLT ALDQDLLLLK ATSAATLSCL GECLNLLQQS VHQAGQPSQK PGASENILGW HGSKSHSTEQ LKNGTLGSLP SASANITWAI LPNSAEDEQT SQPEPEPNSG SELVLSEDEK SDNEDKEETE LGVMEDQRSI I; Positive Samples: A-431, HT-29, A-549, Rat brain; Cellular Location: Cytoplasm, cytoskeleton.

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Like most members, the encoded protein contains an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSBPL10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OSBPL10
Gene Accession No. Q9BXB5, S4R1M9
Gene Alias 4933433D06Rik; C820004B04Rik; FLJ20363; HGNC:16395; OPR-10; ORP10; ORP-10; OSBP9; OSBPL10; OSBP-related protein 10; OTTHUMP00000161257; oxysterol binding protein like 10; oxysterol binding protein-like 10; oxysterol-binding protein-related protein 10; RGD1561652
Gene Symbols OSBPL10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 200-400 of human OSBPL10 (NP_0602542).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114884, 316039, 74486
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.