Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSBPL11 Monoclonal Antibody (OTI2C8), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA501977S


Description
OSBPL11 Monoclonal antibody specifically detects OSBPL11 in Canine, Human, Rat samples. It is validated for Western BlotSpecifications
OSBPL11 | |
Monoclonal | |
0.69 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9BXB4 | |
Mouse | |
Affinity chromatography | |
RUO | |
100856388, 114885, 303888 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Western Blot | |
OTI2C8 | |
Unconjugated | |
OSBPL11 | |
9430097N02Rik; AI132306; AU019795; FLJ13012; FLJ13164; ORP11; ORP-11; OSBP12; OSBPL11; OSBP-related protein 11; oxysterol binding protein like 11; oxysterol binding protein-like 11; oxysterol-binding protein-related protein 11; TCCCIA00292; testicular tissue protein Li 132 | |
Full length human recombinant protein of human OSBPL11 produced in HEK293T cell. | |
30 μL | |
Primary | |
Canine, Human, Rat | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction