Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSBPL2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001332 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001332 Supplier Novus Biologicals Supplier No. NB1001332
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody has been used in 2 publications

OSBPL2 Polyclonal specifically detects OSBPL2 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen OSBPL2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_055650
Gene Alias KIAA0772FLJ20223, ORP-2MGC4307, ORP2MGC8342, OSBP-related protein 2, oxysterol binding protein-like 2, oxysterol-binding protein-like 2, oxysterol-binding protein-related protein 2
Gene Symbols OSBPL2
Host Species Goat
Immunogen Peptide with sequence YFERNFSDCPDIY corresponding to C-Terminus according to NP_055650.1, NP_653081.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 9885
Test Specificity This antibody is expected to recognize both reported isoforms (represented by NP_055650.1 and NP_653081.1)
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.