Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSBPL3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142077
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142077 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142077 Supplier Invitrogen™ Supplier No. PA5142077
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise all reported isoforms (NP_056365.1; NP_663160.1; NP_663161.1; NP_663162.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

ZNF703 belongs to the Elbow/Noc family. It contains 1 C2H2-type zinc finger.ZNF703 may function as a transcriptional repressor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSBPL3
Applications Flow Cytometry, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene OSBPL3
Gene Accession No. Q9H4L5
Gene Alias 1200014M06Rik; 6720421I08Rik; A530055M08; KIAA0704; ORP3; ORP-3; OSBP3; Osbpl3; OSBP-related protein 3; oxysterol binding protein like 3; oxysterol binding protein-like 3; oxysterol-binding protein-related protein 3; RGD1564287
Gene Symbols OSBPL3
Host Species Goat
Immunogen Peptide with sequence MSDEKNLGVSQKL-C (aa 2-14).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26031
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.