Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSBPL9 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB1001336
Description
OSBPL9 Polyclonal specifically detects OSBPL9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
OSBPL9 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FLJ12492, FLJ14629, FLJ14801, FLJ32055, FLJ34384, ORP-9, ORP9MGC15035, OSBP4, OSBP-related protein 9, oxysterol binding protein-like 9, oxysterol-binding protein-related protein 9 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognize all six human isoforms (NP_078862.4; NP_683702.1; NP_683704.2; NP_683705.1; NP_683706.3; NP_683707.3). Reported variants represent identical protein (NP_683702.1; NP_683703.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 2 - 4 ug/ml, Immunohistochemistry-Paraffin 2 - 4 ug/ml, Peptide ELISA Detection limit 1:32000 | |
NP_078862 | |
OSBPL9 | |
Peptide with sequence C-EPLLKRLGAAKH corresponding to C-Terminus according to NP_078862.4, NP_683702.1, NP_683704.2, NP_683705.1, NP_683706.3, NP_683707.3. | |
0.1 mg | |
Lipid and Metabolism | |
114883 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction