Learn More
OSBPL9/ORP9 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304907A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan
This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Multiple transcript variants, most of which encode distinct isoforms, have been identified.Specifications
OSBPL9/ORP9 | |
Polyclonal | |
Unconjugated | |
OSBPL9 | |
OSBPL9; ORP9; FLJ12492; FLJ14629; FLJ14801; FLJ32055; FLJ34384; MGC15035; oxysterol binding protein-like 9; OSBP-related protein 9; oxysterol-binding protein-related protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
114883 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q96SU4 | |
OSBPL9 | |
Between 250 and 300 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.