Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSCAR Antibody (RM0300-11E34), Alexa Fluor™ 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP211836AF350
Description
OSCAR Monoclonal specifically detects OSCAR in Mouse samples. It is validated for Western Blot.Specifications
OSCAR | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
OSCAR | |
Mouse recombinant OSCAR (extracellular domain) | |
0.1 mL | |
Primary | |
Mouse | |
IgG2 |
Western Blot | |
RM0300-11E34 | |
Western Blot | |
hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
Rat | |
Protein A or G purified | |
Immunology | |
126014.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction