Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSCAR Antibody (RM0300-11E34) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$170.97 - $554.59
Specifications
| Antigen | OSCAR |
|---|---|
| Clone | RM0300-11E34 |
| Concentration | LYOPH |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
OSCAR Monoclonal specifically detects OSCAR in Mouse samples. It is validated for Western Blot.Specifications
| OSCAR | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q8VBT3 | |
| 126014 | |
| IgG2 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| RM0300-11E34 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
| OSCAR | |
| Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title