Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSCAR Monoclonal Antibody (5B8), Invitrogen™

Catalog No. PIMA533374
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533374 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533374 Supplier Thermo Scientific Supplier No. MA533374

Rat Monoclonal Antibody

Product is supplied at 1 mL (100 μL/mL).

Recently, a novel member of the leukocyte receptor complex (LRC) encoded family has been identified that is expressed specifically in osteoclast (OC). This protein names OC-associated receptor or OSCAR, play critical roles in the regulation of both innate and adaptive immune responses and bone-specific regulator of OC differentiation. Osteoclasts are multinucleated cells that resorb bone and are essential for bone homeostasis. This protein family plays critical roles in the regulation of both innate and adaptive immune responses. Different from the other LRC members, OSCAR expression is detected specifically in preosteoclasts or mature osteoclasts. OSCAR may be an important bone-specific regulator of osteoclast differentiation. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSCAR
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 5B8
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene OSCAR
Gene Accession No. Q8VBT3
Gene Alias MGC33613; mOSCAR; mOSCAR-M1; mOSCAR-M2; mOSCAR-M3; ocl178; Oscar; osteoclast associated receptor; osteoclast associated receptor OSCAR-S1; osteoclast associated receptor OSCAR-S2; osteoclast associated, immunoglobulin-like receptor; osteoclast-associated immunoglobulin-like receptor; Osteoclast-associated receptor; osteoclast-associated receptor OSCAR; PIGR3; PIgR-3; poly-Ig receptor 3; polymeric immunoglobulin receptor 3
Gene Symbols OSCAR
Host Species Rat
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 232790
Target Species Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.