Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Oscillating Reactions: Yellow to Blue and Back
SCI_ED

Catalog No. s96751
Click to view available options
Class Size:
3 Lab Groups
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.
WARNING: This product can expose you to chemicals including Strong Inorganic Acid Mists containing Sulfuric Acid, which are known to the State of California to cause cancer.

Includes

Material for five demonstrations: 25 mL Hydrogen Peroxide, 12% UN 2984 (3); 25 mL Potassium Iodate, 0.2M solution UN 2796 (3); 75mL Starch solution, 0.03%; and instructions

See the colors change before your eyes.

  • Prepare a flask containing a yellow solution
  • Mix it on a magnetic stirrer
  • Solution turns from yellow to blue and back to yellow

Specifications

Color White
Grade 7 to 14
Age 13 to 20
Includes 3 x 25 mL hydrogen peroxide, 12% un2984, 3 x 25 mL potassium iodate, 0.2 m solution un2796, 1 x 75 mL starch solution, 0.03%
Quantity 1
Dimensions (L x W x H) 5.5 x 4.25 x 6.75 in.
Class Size 3 Lab Groups
Product Type Oscillating Reactions: Yellow to Blue and Back Demonstration Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.