Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113346
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113346 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113346 Supplier Invitrogen™ Supplier No. PA5113346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OSER1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen OSER1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene OSER1
Gene Accession No. Q9D722, Q9NX31
Gene Alias 3230401D17Rik; AI256769; BM-038; C20orf111; dJ1183I21.1; HSPC168; HSPC207; OSER1; Osr1; oxidative stress responsive 1; oxidative stress responsive serine rich 1; oxidative stress responsive serine-rich 1; oxidative stress-responsive 1; oxidative stress-responsive protein 1; Oxidative stress-responsive serine-rich protein 1; Perit1; Peroxide-inducible transcript 1 protein; RGD1303142
Gene Symbols OSER1
Host Species Rabbit
Immunogen Recombinant Human Oxidative stress-responsive serine-rich protein 1 protein (1-292AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51526, 66680
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.