Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSGEP Antibody (OTI9E3), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273163AF647
Description
OSGEP Monoclonal specifically detects OSGEP in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
OSGEP | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
OSGEP | |
Recombinant protein fragment corresponding to amino acids 1-335 of human OSGEP (NP_060277) produced in E.coli. | |
0.1 mL | |
55644.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI9E3 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
EC 3.4.24.57, GCPL1FLJ20411, hOSGEP, KAE1, OSGEP1, O-sialoglycoprotein endopeptidase, probable O-sialoglycoprotein endopeptidase, PRSMG1, t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEP | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat, Canine | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction