Learn More
OSGEP Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI9E3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503002

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Osgep (O-sialoglycoprotein endopeptidase), also known as GCPL1, is a 335 amino acid protein that is a member of the peptidase M22 family. Osgep specifically cleaves the 31-Arg-l-Asp-32 bond in glycophorin A, but it does not cleave desialylated glycoproteins, unglycosylated proteins or glycoproteins that are only N-glycosylated. Though ubiquitously expressed at low levels, highest levels of Osgep are found in liver, skeletal muscle and kidney. OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as three alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine.Specifications
OSGEP | |
Monoclonal | |
Unconjugated | |
OSGEP | |
GAMOS3, GCPL1, KAE1, OSGEP1, PRSMG1, TCS3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
100687555, 290028, 55644, 66246 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI9E3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q8BWU5, Q9NPF4, Q9WVS2 | |
Osgep | |
Human recombit protein fragment corresponding to amino acids 1-335 of human OSGEP produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human, Mouse, Rat | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.