Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSGEPL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117114
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117114 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117114 Supplier Invitrogen™ Supplier No. PA5117114
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse heart, Mouse kidney, Mouse liver, Mouse testis, Rat liver, Rat brain Immunogen sequence: MLILTKTAGV FFKPSKRKVY EFLRSFNFHP GTLFLHKIVL GIETSCDDTA AAVVDETGNV LGEAIHSQTE VHLKTGGIVP PAAQQLHREN IQRIVQEALS ASGVSPSDLS AIATTIKPGL ALSLGVGLSF SLQLVGQLKK PFIPIHHMEA HALTIRLTNK VEFPFLVLLI SGGHCLLALV QGVSDFLLLG KSLDIAPGDM LDKVARRLSL IKHPECSTMS GGKAIEHLAK QGNRFHFDIK PPLHHAKNCD FSFTGLQHVT DKIIMKKEKE.

OSGEPL1, also known as Qri7, is a 414 amino acid protein that belongs to the KAE1/YgjD family and exists as 3 alternatively spliced isoforms. In tRNAs that have codons beginning with adenine, OSGEPL1 is required for the formation of a threonylcarbamoyl group on adenosine. The gene that encodes OSGEPL1 contains about 16,568 bases and maps to human chromosome 2q32.2. Consisting of 237 million bases, chromosome 2 encodes over 1,400 genes and makes up approximately 8% of the human genome. A number of genetic diseases are linked to genes on chromosome 2. Harlequin icthyosis, a rare and morbid skin deformity, is associated with mutations in the ABCA12 gene. The lipid metabolic disorder sitosterolemia is associated with ABCG5 and ABCG8. An extremely rare recessive genetic disorder, Alstrom syndrome, is due to mutations in the ALMS1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSGEPL1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OSGEPL1
Gene Accession No. Q4V7F3, Q6PEB4, Q9H4B0
Gene Alias 2610001M19Rik; AA416452; GCP1; HAMAP-Rule:MF_03179}; N6-L-threonylcarbamoyladenine synthase; OSGEPL1; O-sialoglycoprotein endopeptidase like 1; O-sialoglycoprotein endopeptidase-like 1; O-sialoglycoprotein endopeptidase-like protein 1; O-sialoglycoprotein endopeptidase-like protein 1 {ECO:0000255; probable O-sialoglycoprotein endopeptidase 2; probable tRNA N6-adenosine threonylcarbamoyltransferase, mitochondrial; probable tRNA N6-adenosine threonylcarbamoyltransferase, mitochondrial {ECO:0000255; probable tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1; putative sialoglycoprotease type 2; Qri7; t(6)A synthase; t(6)A37 threonylcarbamoyladenosine biosynthesis protein OSGEPL1; t(6)A37 threonylcarbamoyladenosine biosynthesis protein Osgepl1 {ECO:0000255; tRNA threonylcarbamoyladenosine biosynthesis protein OSGEPL1; tRNA threonylcarbamoyladenosine biosynthesis protein Osgepl1 {ECO:0000255
Gene Symbols OSGEPL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human OSGEPL1 (NP_0717482).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314548, 64172, 72085
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.