Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSGIN1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104338
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104338 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104338 Supplier Invitrogen™ Supplier No. PA5104338
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OSGI1.

OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSGIN1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Osgin1
Gene Accession No. Q9UJX0
Gene Alias 1700012B18Rik; BDGI; BMSC-derived growth inhibitor; Bone marrow stromal cell-derived growth inhibitor; huOKL38; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 1; Okl38; OSGIN1; Ovary, kidney and liver protein 38; oxidative stress induced growth inhibitor 1; oxidative stress-induced growth inhibitor 1; pregnancy-induced growth inhibitor; pregnancy-induced growth inhibitor OKL38; Unknown (protein for MGC:137469)
Gene Symbols Osgin1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OSGIN1(Accession Q9UJX0), corresponding to amino acid residues 417-431.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29948, 71839
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.