Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSGIN1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986751
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6751 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6751 Supplier Bioss Supplier No. BS5723R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

OSGIN1 regulates the differentiation and proliferation of normal cells through the regulation of cell death.

Specifications

Antigen OSGIN1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Osgin1
Gene Accession No. Q9UJX0
Gene Alias 1700012B18Rik; BDGI; BMSC-derived growth inhibitor; Bone marrow stromal cell-derived growth inhibitor; BOS_16987; huOKL38; OKL38; OSGIN1; Ovary, kidney and liver protein 38; oxidative stress induced growth inhibitor 1; Oxidative stress-induced growth inhibitor 1; pregnancy-induced growth inhibitor; Pregnancy-induced growth inhibitor OKL38
Gene Symbols Osgin1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OSGIN1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29948
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.