Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSGIN2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | OSGIN2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
OSGIN2 Polyclonal specifically detects OSGIN2 in Human samples. It is validated for Western Blot.Specifications
| OSGIN2 | |
| Polyclonal | |
| Rabbit | |
| Cell Cycle and Replication | |
| hT41C8orf1chromosome 8 open reading frame 1, oxidative stress induced growth inhibitor family member 2, oxidative stress-induced growth inhibitor 2 | |
| OSGIN2 | |
| IgG | |
| 56 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q9Y236 | |
| 734 | |
| Synthetic peptides corresponding to OSGIN2 (oxidative stress induced growth inhibitor family member 2) The peptide sequence was selected from the C terminal of OSGIN2. Peptide sequence ECIKEANLFALGPLVGDNFVRFLKGGALGVTRCLATRQKKKHLFVERGGG. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title