Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSGIN2 Antibody (OTI7D12), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273164F
Description
OSGIN2 Monoclonal specifically detects OSGIN2 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
OSGIN2 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
OSGIN2 | |
Recombinant protein fragment corresponding to amino acids 1-250 of human OSGIN2 (NP_004328) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Immunohistochemistry | |
OTI7D12 | |
Immunohistochemistry | |
hT41C8orf1chromosome 8 open reading frame 1, oxidative stress induced growth inhibitor family member 2, oxidative stress-induced growth inhibitor 2 | |
Mouse | |
Affinity Purified | |
Cell Cycle and Replication | |
734.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction