Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSGIN2 Monoclonal Antibody (OTI6E10)
GREENER_CHOICE

Catalog No. PIMA527079
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527079 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527079 Supplier Invitrogen™ Supplier No. MA527079
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OSGIN2 Monoclonal Antibody for Western Blot, IHC (P)

May be involved in meiosis or the maturation of germ cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSGIN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6E10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OSGIN2
Gene Accession No. Q9Y236
Gene Alias BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323
Gene Symbols OSGIN2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-250 of OSGIN2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 734
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.