Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSGIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565627
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565627 Supplier Invitrogen™ Supplier No. PA565627
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EGSMLTISFGS WMELPGLKFK DWVSSKRRSL KGDRVMPEEI ARYYKHYVKV MGLQKNFREN TYITSVSRLY RDQDDDDIQD RDISTKHLQ Highest antigen sequence identity to the following orthologs - mouse 91%, rat 91%.

May be involved in meiosis or the maturation of germ cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSGIN2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OSGIN2
Gene Accession No. Q9Y236
Gene Alias BC010311; C230027H09Rik; C8orf1; hT41; LOW QUALITY PROTEIN: oxidative stress-induced growth inhibitor 2; OSGIN2; oxidative stress induced growth inhibitor family member 2; oxidative stress-induced growth inhibitor 2; RGD1306323
Gene Symbols OSGIN2
Host Species Rabbit
Immunogen Recombinant Human OSGIN2. Recombinant protein control fragment (Product #RP-106294).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 734
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.