Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSM™ All-Natural Glass and Surface Cleaner

Catalog No. 15336512
Click to view available options
Volume (English):
1 gal.

Catalog No. 15-336-512

Supplier: OSM™ 81008201238

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Streak-free shine with no smearing

Ideal for a variety of surfaces in the bathroom, kitchen, and work areas. Use on glass, plastic, rubber, vinyl, labware, and most metals. Renewable plant-derived formula free of phosphates, glycol ether, phthalates, and ammonia.

  • Perfect for glass test tubes, Petri dishes, and weigh plates
  • Water-based product with natural cleaning agents and plant-based surfactants
  • Environmentally friendly alternative to harsh chemicals; safe to use around people and animals
  • Disposes down drain
  • No ammonia, no harsh chemical fumes or vapors

Specifications

Form Liquid
Appearance Clear
Boiling Point >100°C
For Use With (Application) Use in lab, on lab Glassware, kitchen, bathroom or wherever Glass is present
Volume (English) 1 gal.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.