Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSM™ All-Natural No-Rinse Floor Cleaner

Catalog No. p-3556864
Click to view available options
Volume (English):
1 gal.
This item is not returnable. View return policy
This item is not returnable. View return policy

Cleans gently, leaving a beautiful luster; safe for use around people and animals

Perfect for cabinets, molding, doors, painted surfaces, and more.

  • Use on wood, vinyl, laminate, and tile
  • Penetrates and emulsifies grease, dirt, and grime
  • Highly effective cleaning agents with plant-based surfactants leave no harmful residue
  • Renewable plant-derived formula free of phosphates, glycol ether, and phthalates
  • Ozone-safe formula disposes down the drain
  • No harsh chemical fumes or vapor

Specifications

Product Type Floor Cleaner
Form Liquid
Appearance Green
For Use With (Application) For use on tile, laminate, Vinyl, Wood and painted surfaces, safe for cleaning cabinets, molding, paneling and doors
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.