Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human OSMR beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $479.50
Specifications
Antigen | OSMR beta |
---|---|
Clone | 469229 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB43891SP
![]() |
R&D Systems
MAB43891SP |
25 μg |
Each for $200.50
|
|
|||||
MAB43891100
![]() |
R&D Systems
MAB43891100 |
100 μg |
Each for $479.50
|
|
|||||
Description
OSMR beta Monoclonal specifically detects OSMR beta in Human samples. It is validated for ELISA.Specifications
OSMR beta | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q99650 | |
9180.0 | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
469229 | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
Mouse myeloma cell line NS0-derived recombinant human OSM R beta , Glu28-Ser739, Accession No. Q99650 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human OSM R beta in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title