Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSMR beta Rat anti-Mouse, Alexa Fluor 405, Clone: 118125, R&D Systems™

Catalog No. FAB662V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB662V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB662V100 Supplier R&D Systems Supplier No. FAB662V100UG
This item is not returnable. View return policy

Rat Monoclonal Antibody

OSMR beta Monoclonal antibody specifically detects OSMR beta in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen OSMR beta
Applications Flow Cytometry
Classification Monoclonal
Clone 118125
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 405
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O70458
Gene Alias IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSM R beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1
Gene Symbols Osmr
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse OSM R beta Glu24-Leu738 Accession # O70458
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Test Specificity Detects mouse OSM R beta in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CLC, rhCNTF, rhOSM, recombinant mouse (rm) CT-1, rmIL-6, rmIL-11, or rmLIF is observed.
Target Species Mouse
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.