Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Osmic Acid Anhydride (Crystalline/Laboratory), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
0.25 g
1 g
Packaging:
Ampule
Specifications
Specifications
| Boiling Point | 130°C |
| Melting Point | 39.5°C |
| Color | White |
| Physical Form | Solid |
| Quantity | 1 g |
| Formula Weight | 254.23 |
| Grade | Laboratory |
| Identification | Pass Test |
| Packaging | Ampule |
| Vapor Pressure | 7mmHg at 20°C |
| Show More |
Safety and Handling
| DOTInformation | DOT Class 6.1, : Poison |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction