Learn More
Osmium tetroxide, 2.5 wt.% solution in tert-Butanol, stabilized, Thermo Scientific Chemicals
$50.25 - $446.65
Specifications
Chemical Name or Material | Osmium tetroxide |
---|---|
CAS | 20816-12-0, 75-65-0 |
Color | White to Yellow |
Physical Form | Crystals, Fused Mass or Solution |
Molecular Formula | O4Os |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC197450010
|
Thermo Scientific Chemicals
197450010 |
1 mL | Glass bottle |
Each for $50.25
Order in Bulk
Request bulk or custom formats |
|
||||
AC197450050
|
Thermo Scientific Chemicals
197450050 |
5 mL | Glass bottle |
Each
for $120.65
Save 23% Reg :
Order in Bulk
Request bulk or custom formats |
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
AC197450250
|
Thermo Scientific Chemicals
197450250 |
25 mL | Glass bottle |
Each
for $446.65
Save 15% Reg :
Order in Bulk
Request bulk or custom formats |
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentSpecifications
Osmium tetroxide | |
White to Yellow | |
O4Os | |
MFCD00011150 | |
4°C | |
osmium tetroxide, osmium tetraoxide, osmic acid, perosmic oxide, osmic acid anhydride, osmium viii oxide, oso4, perosmic acid anhydride, osmiumtetroxide, osmium oxide, t-4 | |
O=[Os](=O)(=O)=O | |
254.23 | |
254.2 | |
Glass bottle | |
0.8110g/mL |
20816-12-0, 75-65-0 | |
Crystals, Fused Mass or Solution | |
OsO4 | |
01,759; 02,301; 04,361; 05,141; 06,424; 10,290; 12,358; 13,186; 14,235; 15,240; 16,249; 17,236 | |
15, 6990 | |
VUVGYHUDAICLFK-UHFFFAOYSA-N | |
tetraoxoosmium | |
30318 | |
2.5 wt% Min. | |
0.811 |
Safety and Handling
GHS H Statement
Toxic if inhaled.
Harmful if swallowed.
Fatal in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause allergy or asthma symptoms o
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
GHS Signal Word: Danger
EINECSNumber : 244-058-7
RTECSNumber : RN1140000
TSCA : TSCA
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.