Learn More
American Educational Products Osmosis Apparatus Kit
SCI_ED
Catalog No. S41305 Shop All American Educational Products LLC Products
Change view
Click to view available options
Osmosis Apparatus W/jar
Catalog No.
S41305 Osmosis Apparatus W/jar
1 options
Catalog No. S41305 Supplier American Educational Products Supplier No. 715017
Includes
  • Double thistle tube
  • Battery jar
  • Metal holder
  • Instructions
Requires
Sugar solution
Observe a dramatic demonstration of the principle of osmosis.
Consists of a 40cm long, double thistle tube; one end is covered with an animal membrane. Add sugar solution to the top of the tube — the liquid rises in the tube as it moves through the membrane.
Specifications
Product Type Double Thistle Osmosis Apparatus
For Use With (Application) For illustrating the principles of osmosis
Includes Instruction manual, battery jar, thistle tube with membrane and a stable support