Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSMR beta Antibody (007), DyLight 594 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289851DL594
Description
OSMR beta Monoclonal antibody specifically detects OSMR beta in Human samples. It is validated for ELISASpecifications
OSMR beta | |
Monoclonal | |
DyLight 594 | |
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human OSMR beta (NP_003990.1; Met 1-Met 740). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
007 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
9180 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction