Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSMR beta Antibody (118125) [Allophycocyanin], Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB662A
Description
OSMR beta Monoclonal antibody specifically detects OSMR beta in Mouse samples. It is validated for Western Blot, ImmunocytochemistrySpecifications
OSMR beta | |
Monoclonal | |
APC | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9180 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Immunocytochemistry | |
118125 | |
Western Blot, Immunocytochemistry | |
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
Mouse myeloma cell line NS0-derived recombinant mouse OSM R beta , Glu24-Leu738, Accession # O70458 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction