Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSMR beta Antibody (202), Allophycocyanin, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290615APC
Description
OSMR beta Monoclonal antibody specifically detects OSMR beta in Mouse samples. It is validated for ELISASpecifications
OSMR beta | |
Monoclonal | |
APC | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
9180 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
202 | |
ELISA | |
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
Rabbit immunized with purified, recombinant Mouse OSMR beta (Uniprot#: O70458-1, Met1-Leu738) | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction