Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSMR beta Rabbit anti-Human, Clone: 2, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | OSMR beta |
---|---|
Clone | 2 |
Dilution | Flow Cytometry 1:25-1:100, Sandwich ELISA Capture 1:250-1:2000 |
Applications | Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
OSMR beta Monoclonal specifically detects OSMR beta in Human samples. It is validated for Flow Cytometry, Sandwich ELISA Capture.Specifications
OSMR beta | |
Flow Cytometry 1:25-1:100, Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
9180 | |
IgG | |
Protein A purified |
2 | |
Flow Cytometry, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
IL-31 receptor subunit beta, IL-31R subunit beta, IL-31RB, IL-31R-beta, Interleukin-31 receptor subunit beta, MGC150627, MGC75127, oncostatin M receptor, oncostatin-M specific receptor beta subunit, oncostatin-M-specific receptor subunit beta, OSMR, OSMRB, OSMRBMGC150626, PLCA1 | |
Recombinant Human OSMR beta protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title