Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSMR Recombinant Rabbit Monoclonal Antibody (002), APC
GREENER_CHOICE

Catalog No. PIMA546728
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546728 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA546728 Supplier Invitrogen™ Supplier No. MA546728
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human OSMR (Met1-Met740) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF.Oncostatin M is a member of the IL6 family of cytokines. Functional receptors for IL6 family cytokines are multisubunit complexes involving members of the hematopoietin receptor superfamily. Many IL6 cytokines utilize gp130 as a common receptor subunit. OSM binds to the gp130 receptor subunit and, in association with the leukemia inhibitory factor receptor, induces a proliferative response in permissive cells. OSMR is an alternative subunit (for an OSM receptor complex (a heterodimer of gp130 and OSMR) that is activated by OSM but not by LIF. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSMR
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone 002
Concentration 100 μg/mL
Conjugate APC
Formulation PBS with 0.5% BSA and 0.03% ProClin 300
Gene OSMR
Gene Accession No. Q99650
Gene Alias IL-31 receptor subunit beta; IL-31R subunit beta; IL-31RB; IL-31R-beta; interleukin-31 receptor subunit beta; MGC150626; MGC150627; MGC75127; oncostatin M receptor; oncostatin receptor; oncostatin-M specific receptor beta subunit; oncostatin-M specific receptor subunit beta; oncostatin-M-specific receptor subunit beta; OSMR; Osmrb; PLCA1
Gene Symbols OSMR
Host Species Rabbit
Immunogen Recombinant protein ecoding Human OSMR (Met1-Met740).
Purification Method Protein A
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9180
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.