Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sekisui OSOM Flu SARS-CoV-2 Combo Test Kit

Catalog No. 22321143 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Detection Method:
Manual Antigen
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detection Method
22-321-143 Manual Antigen
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-143 Supplier Sekisui Supplier No. 1080
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

OSOM Flu SARS-CoV-2 Combo Test is a lateral flow in vitro diagnostic for rapid and simultaneous detection and differentiation of influenza A, influenza B, and SARS-CoV-2 antigens for Healthcare providers. For Emergency Use Authorization Only.

  • One Sample, One Test, and Three Steps to reliably differentiate between SARS-CoV-2, Flu A, and Flu B.
  • Improve clinical outcomes by quickly differentiating between SARS-CoV-2, Flu A, and Flu B in 10 minutes with one sample collection.
  • The signature OSOM™ QC Inside™ feature gives you two additional tests in each box so that external quality controls can be performed without impacting your bottom line.
  • The OSOM™ Flu SARS-CoV-2 Combo Test Kit is EUA approved for anterior nasal swab samples in the CLIA waived, moderate, and high complexity laboratory settings.
TRUSTED_SUSTAINABILITY

Specifications

CE Marker No
Clia Complexity Waived (EUA)
Detectable Analytes SARS-CoV-2, Influenza A, Influenza B
Detection Method Manual Antigen
Format Kit
Sample Type Anterior Nasal Swab
Sensitivity High
Type lateral flow immunochromatographic
For Use With (Application) Detection and Differentiaion of influenza A, influenza B, and SARS-CoV-2
Form Ready-to-Use (RTU)

In the USA, this product has not been FDA cleared or approved, but has been authorized by FDA under an Emergency Use Authorization. This product has been authorized only for the detection of proteins from SARS-CoV-2, influenza A and influenza B, not for any other viruses or pathogens. The emergency use of this product is only authorized for the duration of the declaration that circumstances exist justifying the authorization of emergency use of in vitro diagnostics for detection and/or diagnosis of COVID-19 under Section 564(b)(1) of the Federal Food, Drug, and Cosmetic Act, 21 U.S.C. §360bbb-3(b)(1), unless the declaration is terminated or authorization is revoked sooner.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.