Learn More
SEKISUI Diagnostics™ OSOM™ iFOB Test
Shop All SEKISUI Diagnostics ProductsDescription
Includes
- 25-OSOM iFOB Test device in a sealed foil pouch
- 25 Stool collection tube, with sampler, containing extraction buffer solution (PBS)
- 25 Collection paper
- 25 Mailing Envelope
- Instructions for use
- 25 Patient instructions for use
- 25 Biohazard labeled bags with absorbent pads
- Two extra test devices for external QC testing
OSOM iFOBT Control Kit (Cat. No. 22-321-112)
1mL OSOM iFOBT Control Positive Control and 1mL OSOM iFOBT Control Negative Control
OSOM iFOBT Patient Mailers (Cat. No. 22-321-110)
25 Mailers
OSOM iFOBT Stool Collection Tubes (Cat. No. 22-321-111)
25 Collection tubes
The OSOM™ iFOB Test is a rapid qualitative test for the immunochemical detection of fecal occult blood/human hemoglobin (hHb) in human fecal specimens as an aid in the diagnosis of gastrointestinal disorders such as: diverticulitis, colitis, polyps, and colorectal cancer. Convenient take home kit, patients can collect in the privacy of their own home to increase compliance in screening.
- Aids in prevention and helps reduce the risk of developing more serious ulcers and other health issues
- Lower healthcare costs - early diagnosis leads to the reduction of cancer treatment expenses, hospital stays, and long-term care costs
- Improved patient compliance - no dietary or drug restrictions, with an easy-to-use at home sample collection kit and procedure
- Quick turnaround - obtain results in 5 to 10 minutes with less than 1 minute of hands-on time
- Trust your results - highly specific to human hemoglobin with analytical sensitivity of 50 ng hHb/mL buffer or 50 μg hHb/g feces
- QC Inside™ - two extra tests included at no additional charge to run external quality control testing for more confidence in your results
Recommended Storage
Store the kit or kit contents 2° to 30°C until use. Do not freeze.

Specifications
Specifications
Content And Storage | Store the kit or kit contents 2°C to 30°C (35.6°F to 86°F) until use. Do not freeze. |
CPT Code | Diagnostic 82274QW; Screening G0328QW |
Clia Complexity | Waived |
Detectable Analytes | Hemoglobin |
Format | Cassette |
Sample Type | Stool |
Sensitivity | 50 ng hHb/mL Buffer or 50 μg hHb/g Feces |
Type | Rapid Test Kits |
For Use With (Application) | Fecal Occult Blood Detection |
Includes | 25 OSOM iFOB Test device in a sealed foil pouch, 25 Stool collection tube, with sampler, containing extraction buffer solution (PBS), 25 Collection paper, 25 Mailing Envelopes, Instructions for use, 25 Patient instructions for use, 25 Biohazard labeled bags with absorbent pads, Two extra test devices for external QC testing |
Show More |
For in vitro diagnostic use only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.