Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SEKISUI Diagnostics™ OSOM™ Ultra Plus Flu A and B Test Kit

Catalog No. 22321130 Shop All SEKISUI Diagnostics Products
Change view
Click to view available options
Format:
Manual Dipstick
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
22-321-130 Manual Dipstick
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-321-130 Supplier SEKISUI Diagnostics™ Supplier No. 1032
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Use OSOM™ Ultra Plus Flu A and B Test Kit to detect influenza type A and B in just 10 minutes with minimal hands-on time, simplified workflow, and two additional tests for quality testing at no additional charge.

The OSOM™ Ultra Plus Flu A and B Test is an in vitro rapid qualitative test that detects influenza type A and B nucleoprotein antigens directly from nasal and nasopharyngeal swab specimens obtained from patients. High sensitivity and specificity allow for accurate diagnostic testing that is both efficient and cost-effective, allowing for diagnosis and treatment during the same visit.

  • Fast diagnosis - read test results in just 10 minutes
  • Trust your results - high sensitivity and specificity for more accurate diagnosis
  • Save time - pre-filled extraction buffer vial to reduce hands-on time
  • Ready-to-use - everything needed to run test is included, with no need for specialized instruments
  • QC Inside™ - two extra tests included at no additional charge to run external quality control testing for more confidence in your results
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CLIA waived, 510k approved
Content And Storage Room Temperature (15°C to 30°C, 59°F to 86°F)
Control Sets Included
CPT Code 87804QW (Flu A), 87804QW-59 (Flu B)
Clia Complexity Waived (Nasal and NP Swab)
Detectable Analytes Influenza A and B
Detection Method Influenza antigens from nasal or NP swabs
Format Manual Dipstick
Sensitivity A: 90.3% B: 88.0%
Type Rapid Test
Includes 25 test sticks, 25 nasal swabs, 25 extraction buffer vials, Influenza A+ Control Swab, Influenza B+ Control Swab, IFU, QRG, Workstation
Packaging Type Test Kit
Species Performance characteristics for influenza A were established during the U.S. 2018-2019 influenza season when A/H1N1pdm09 and influenza A/H3N2 were the predominant influenza A viruses in circulation, and the influenza B Yamagata and Victoria lineages were in co-circulation
Specificity A: 96.7% B: 99.2%
Test Time 10 min.
Analysis Time 10 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.