Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OspC Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP221683
Description
OspC Polyclonal specifically detects OspC in Bacteria samples. It is validated for Western Blot.Specifications
OspC | |
Polyclonal | |
Unconjugated | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
ospC, BB_B19, Outer surface protein C, pc | |
Rabbit | |
22.34 kDa | |
0.1 mg | |
Primary | |
Cross-adsorbed against MBP from monospecific antiserum by chromatography. This antibody is specific for Borrelia burgdorferi OspC protein. | |
Bacteria | |
Purified |
Western Blot, ELISA | |
LYOPH | |
Western Blot 1:1000 | |
Q07337 | |
BB_B19 | |
MBP-fusion protein corresponding to Borrelia burgdorferi OspC protein. (Uniprot: Q07337) | |
Protein A purified | |
RUO | |
1194415 | |
Reconstitute with 100 ul deionized water (or equivalent) | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction