Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OspC Polyclonal Antibody, Rockland™
SDP

Catalog No. 501057638
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-105-7638 25 μL
50-194-3808 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-105-7638 Supplier Rockland Immunochemicals Supplier No. 200401C11S
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to restoration. For extended storage aliquot contents and freeze at -20°C or below. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for Borrelia burgdorferi OspC protein. A BLAST analysis was used to suggest cross-reactivity with p39 from B. burgdorferi, afzelii, and valaisiana sources based on 100% homology with the immunizing sequence. Partial reactivity is expected against B. japonica and americana sources based on 94% homology. Cross-reactivity with OspC from other sources has not been determined.

Outer Surface Protein C, or OspC, is a 20.7 kDa immunogenic protein on the outer surface of the spirochete Borrelia burgdorferi. Its function is not known, but it is located with lipid-anchoring sites on the outer cell membrane.

Specifications

Antigen OspC
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.95 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen MBP-fusion protein corresponding to Borrelia burgdorferi OspC protein.
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.