Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSR1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501572295
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-2295 100 μL
50-157-2296 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-157-2295 Supplier Bethyl Laboratories, Inc Supplier No. IHC00552
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine, Dog, Horse, Guinea pig_10141, Orangutan, Monkey, Gorilla, Chimpanzee, Chine

Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.

Specifications

Antigen OSR1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene OXSR1
Gene Accession No. O95747
Gene Alias OTTHUMP00000209062; oxidative stress-responsive 1 protein; serine/threonine-protein kinase OSR1
Gene Symbols OXSR1
Host Species Rabbit
Immunogen Between 375 and 425
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9943
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.