Learn More
OSR1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301579AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Mouse and Orangutan
Regulates downstream kinases in response to environmental stress. May also have a function in regulating the actin cytoskeleton.Specifications
| OSR1 | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O95747 | |
| OXSR1 | |
| Between 375 and 425 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| OXSR1 | |
| OTTHUMP00000209062; oxidative stress-responsive 1 protein; serine/threonine-protein kinase OSR1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9943 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.