Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116805
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116805 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116805 Supplier Invitrogen™ Supplier No. PA5116805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: SKOV3, 293T, Mouse testis, Mouse pancreas, Rat large intestine, Rat kidney Immunogen sequence: HMQESPHKCP TCGRTFNQRS NLKTHLLTHT DIKPYSCEQC GKVFRRNCDL RRHSLTHTPR QDF.

OSR2 is a mammalian homolog of the Drosophila odd-skipped family of transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSR2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.91 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene OSR2
Gene Accession No. Q6AY34, Q8N2R0, Q91ZD1
Gene Alias 5430409I15Rik; odd-skipped related 2; odd-skipped related transciption factor 2; Osr2; Osr2A; Osr2B; Protein odd-skipped-related 2
Gene Symbols OSR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-312 of human OSR2 (NP_0011359341).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 107587, 116039, 315039
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.