Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSSA Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories

Catalog No. 501566333
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6333 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6333 Supplier Bethyl Laboratories, Inc Supplier No. A303889AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

FAM120A (C9orf10) is a member of the constitutive coactivator of PPAR -gamma family and the gene was mapped to chromosome 9q22.31. FAM120A was recently detected within the Pur-alpha-containing mRNA-protein complex in the brain. As a novel RNA-binding protein, FAM120A is a critical component of the oxidative stress-induced survival signaling. It may participate in mRNA transport in the cytoplasm. FAM120A activates src family kinases and acts as a scaffolding protein enabling src family kinases to phosphorylate and activate PI3-kinase. FAM120A protects cells from apoptosis through activation of SRKs in response to oxidative stress. Blocking of the survival signaling mediated by FAM120A, which sensitizes the cancer cells to stress-induced apoptosis, may be a novel therapeutic approach for gastric scirrhous carcinoma cells.

Specifications

Antigen OSSA
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene FAM120A
Gene Accession No. Q6A0A9, Q9NZB2
Gene Alias C9orf10, DNAPTP1, DNAPTP5, OSSA; DNA polymerase-transactivated protein 1; DNA polymerase-transactivated protein 5; constitutive coactivator of PPAR-gamma-like protein 1; oxidative stress-associated Src activator
Gene Symbols Fam120a
Host Species Rabbit
Immunogen Between 1068 and 1118
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218236, 23196
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.