Learn More
Invitrogen™ OST 577 Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA560169

Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
OST 577 is a biosimilar that targets HBV.
Specifications
OST 577 | |
Recombinant Monoclonal | |
Unconjugated | |
Human | |
100 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
1 mg/mL | |
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.