Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OSTalpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP157085
Description
OSTalpha Polyclonal specifically detects OSTalpha in Human, Rat samples. It is validated for Western Blot.Specifications
OSTalpha | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
MGC39807, organic solute transporter alpha, organic solute transporter subunit alpha, OSTA, OST-alpha | |
Rabbit | |
Affinity purified | |
RUO | |
200931 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q86UW1 | |
SLC51A | |
Synthetic peptides corresponding to OSTalpha (organic solute transporter alpha) The peptide sequence was selected from the middle region of OSTalpha. Peptide sequence LLMLGPFQYAFLKITLTLVGLFLVPDGIYDPADISEGSTALWINTFLGVS. | |
100 μL | |
Primary | |
This product is specific to Subunit or Isoform: alpha. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction