Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OSTC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534060
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534060 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534060 Supplier Invitrogen™ Supplier No. PA534060
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset (100%) Mouse, Rat, Hamster, Panda, Bovine, Dog, Pig, Opossum, Xenopus (94%).

OSTC is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OSTC
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene OSTC
Gene Accession No. Q9NRP0
Gene Alias 2310008M10Rik; 5730557H03Rik; DC2; HDCMD45P; HSPC307; Hydrophobic protein HSF-28; oligosaccharyltransferase complex non-catalytic subunit; oligosaccharyltransferase complex subunit; oligosaccharyltransferase complex subunit (non-catalytic); oligosaccharyltransferase complex subunit OSTC; Ostc; ostc {ECO:0000250; RGD1560708; UniProtKB:Q9NRP0}
Gene Symbols OSTC
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from N-Terminus of human OSTC.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58505
Target Species Human, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.